Learn More
- Home
- Products
- Microbiology Quality Control Supplies
- Salmonella Antisera
- BD Difco™ Salmonella O Antisera and Quality Control Antigens
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Product Code. 17322361
Change view
Click to view available options
Product Type:
Antiserum
Unit Size:
3mL
Each
| Product Code. | Product Type | unitSize |
|---|---|---|
| 17322361 | Antiserum | Each |
| 11768153 | Antiserum | 3mL |
| 11788153 | Antiserum | 3mL |
| 11758153 | Antiserum | 3mL |
| 17312361 | Antiserum | Each |
| 11742804 | Antiserum | 3mL |
| 11792844 | Antiserum | 3mL |
| 11722804 | Antiserum | 3mL |
| 11782904 | Antiserum | 3mL |
| 11792734 | Antiserum | 3mL |
| 11702744 | Antiserum | 3mL |
| 11712744 | Antiserum | 3mL |
| 17302361 | Antiserum | Each |
| 11748153 | Antiserum | 3mL |
| 11732814 | Antiserum | 3mL |
| 11772804 | Antiserum | 3mL |
| 11742814 | Antiserum | 3mL |
| 11752814 | Antiserum | 3mL |
| 11762814 | Antiserum | 3mL |
| 11742844 | Antiserum | 3mL |
| 11702854 | Antiserum | 3mL |
| 11772844 | Antiserum | 3mL |
| 11752844 | Antiserum | 3mL |
| 11782844 | Antiserum | 3mL |
| 11752854 | Antiserum | 3mL |
| 11742854 | Antiserum | 3mL |
| 11772854 | Antiserum | 3mL |
| 11792874 | Antiserum | 3mL |
| 11702884 | Antiserum | 3mL |
| 11722884 | Antiserum | 3mL |
| 11732884 | Antiserum | 3mL |
| 11792904 | Antiserum | 3mL |
| 17342361 | Antiserum | Each |
This item is not returnable.
View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
This item is not returnable.
View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
- Lyophilized, polyclonal rabbit antisera
- Polyvalent, specific for somatic antigens
- Cross-reactions are possible because serogroups may share non-major group antigens
- When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
- For in vitro diagnostic use
- containing approximately 0.2% sodium azide as a preservative
Specifications
Specifications
| Certifications/Compliance | For in vitro diagnostic use |
| Product Type | Antiserum |