Learn More
- Home
- Products
- Microbiology Quality Control Supplies
- Salmonella Antisera
- BD Difco™ Salmonella O Antisera and Quality Control Antigens
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Product Code. 11788153| Product Code. | Product Type | unitSize |
|---|---|---|
| 11788153 | Antiserum | 3mL |
| 11768153 | Antiserum | 3mL |
| 11758153 | Antiserum | 3mL |
| 17312361 | Antiserum | Each |
| 17322361 | Antiserum | Each |
| 11742804 | Antiserum | 3mL |
| 11792844 | Antiserum | 3mL |
| 11722804 | Antiserum | 3mL |
| 11782904 | Antiserum | 3mL |
| 11792734 | Antiserum | 3mL |
| 11702744 | Antiserum | 3mL |
| 11712744 | Antiserum | 3mL |
| 17302361 | Antiserum | Each |
| 11748153 | Antiserum | 3mL |
| 11732814 | Antiserum | 3mL |
| 11772804 | Antiserum | 3mL |
| 11742814 | Antiserum | 3mL |
| 11752814 | Antiserum | 3mL |
| 11762814 | Antiserum | 3mL |
| 11742844 | Antiserum | 3mL |
| 11702854 | Antiserum | 3mL |
| 11772844 | Antiserum | 3mL |
| 11752844 | Antiserum | 3mL |
| 11782844 | Antiserum | 3mL |
| 11752854 | Antiserum | 3mL |
| 11742854 | Antiserum | 3mL |
| 11772854 | Antiserum | 3mL |
| 11792874 | Antiserum | 3mL |
| 11702884 | Antiserum | 3mL |
| 11722884 | Antiserum | 3mL |
| 11732884 | Antiserum | 3mL |
| 11792904 | Antiserum | 3mL |
| 17342361 | Antiserum | Each |
Product Code. 11788153
Brand: BD 211783
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
- Lyophilized, polyclonal rabbit antisera
- Polyvalent, specific for somatic antigens
- Cross-reactions are possible because serogroups may share non-major group antigens
- When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
- For in vitro diagnostic use
- containing approximately 0.2% sodium azide as a preservative
Specifications
Specifications
| Certifications/Compliance | For in vitro diagnostic use |
| Product Type | Antiserum |