Learn More
- Home
- Products
- Microbiology Quality Control Supplies
- Salmonella Antisera
- BD Difco™ Salmonella O Antisera and Quality Control Antigens
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Product Code. 11768153| Product Code. | Product Type | unitSize |
|---|---|---|
| 11768153 | Salmonella O Antiserum Group M Factor 28 | 3mL |
| 11788153 | Salmonella O Antiserum Group N Factor 30 | 3mL |
| 11758153 | Salmonella O Antiserum Group J Factor 17 | 3mL |
| 17312361 | Salmonella O Antiserum Group G Factors 13, 22 | Each |
| 17322361 | Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25 | Each |
| 11742804 | Salmonella O Antiserum Group O Factor 35 | 3mL |
| 11792844 | Salmonella O Antiserum Factor 14 | 3mL |
| 11722804 | Salmonella O Antiserum Group L Factor 21 | 3mL |
| 11782904 | Salmonella O Antiserum Group D1 Factors 1, 9, 12 | 3mL |
| 11792734 | Salmonella O Antiserum Factor 10, Group E1 | 3mL |
| 11702744 | Salmonella O Antiserum Factor 15, Groups E2 and E3 | 3mL |
| 11712744 | Salmonella O Antiserum Factor 19, Group E4 | 3mL |
| 17302361 | Salmonella O Antiserum Group F Factor 11 | Each |
| 11748153 | Salmonella O Antiserum Factor 34, Group E3 | 3mL |
| 11732814 | Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H | 3mL |
| 11772804 | Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L | 3mL |
| 11742814 | Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O | 3mL |
| 11752814 | Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U | 3mL |
| 11762814 | Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z | 3mL |
| 11742844 | Salmonella O Antiserum Poly F, Groups 51-55 | 3mL |
| 11702854 | Salmonella O Antiserum Factor 20 | 3mL |
| 11772844 | Salmonella O Antiserum Factor 4, Group B | 3mL |
| 11752844 | Salmonella O Antiserum Poly G, Groups 56-61 | 3mL |
| 11782844 | Salmonella O Antiserum Factor 5, Group B | 3mL |
| 11752854 | Salmonella O Antiserum Factor 23, Group G2 | 3mL |
| 11742854 | Salmonella O Antiserum Factor 22, Group G1 | 3mL |
| 11772854 | Salmonella O Antiserum Factor 27 | 3mL |
| 11792874 | Salmonella O Antiserum Factor 2 | 3mL |
| 11702884 | Salmonella O Antiserum Factors 4, 5, Group B | 3mL |
| 11722884 | Salmonella O Antiserum Factor 8, Groups C2 and C3 | 3mL |
| 11732884 | Salmonella O Antiserum Factor 9, Group D | 3mL |
| 11792904 | Salmonella O Antiserum Group E1 Factors 3, 10 | 3mL |
| 17342361 | Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34 | Each |
Product Code. 11768153
Brand: BD 211781
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
- Lyophilized, polyclonal rabbit antisera
- Polyvalent, specific for somatic antigens
- Cross-reactions are possible because serogroups may share non-major group antigens
- When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
- For in vitro diagnostic use
- containing approximately 0.2% sodium azide as a preservative
Specifications
Specifications
| Certifications/Compliance | For in vitro diagnostic use |
| Product Type | Salmonella O Antiserum Group M Factor 28 |