Learn More
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Product Code. 11732814
Change view
Click to view available options
Product Type:
Antiserum
Unit Size:
3mL
Each
Product Code. Product Type unitSize
11732814 Antiserum 3mL
11768153 Antiserum 3mL
11788153 Antiserum 3mL
11758153 Antiserum 3mL
17312361 Antiserum Each
17322361 Antiserum Each
11742804 Antiserum 3mL
11792844 Antiserum 3mL
11722804 Antiserum 3mL
11782904 Antiserum 3mL
11792734 Antiserum 3mL
11702744 Antiserum 3mL
11712744 Antiserum 3mL
17302361 Antiserum Each
11748153 Antiserum 3mL
11772804 Antiserum 3mL
11742814 Antiserum 3mL
11752814 Antiserum 3mL
11762814 Antiserum 3mL
11742844 Antiserum 3mL
11702854 Antiserum 3mL
11772844 Antiserum 3mL
11752844 Antiserum 3mL
11782844 Antiserum 3mL
11752854 Antiserum 3mL
11742854 Antiserum 3mL
11772854 Antiserum 3mL
11792874 Antiserum 3mL
11702884 Antiserum 3mL
11722884 Antiserum 3mL
11732884 Antiserum 3mL
11792904 Antiserum 3mL
17342361 Antiserum Each
33 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code. 11732814
Brand: BD 225351
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
  • Lyophilized, polyclonal rabbit antisera
  • Polyvalent, specific for somatic antigens
  • Cross-reactions are possible because serogroups may share non-major group antigens
  • When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
  • For in vitro diagnostic use
  • containing approximately 0.2% sodium azide as a preservative
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance For in vitro diagnostic use
Product Type Antiserum
compliance-icons
Product Identifier
  • Vial Sal O Antiserum Poly B Lyophilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium Azide