Learn More
BD Difco™ Neisseria Meningitidis Antisera
Product Code. 11758333
Change view
Click to view available options
Quantity:
1 mL
3 mL
Description:
Antiserum Group A
Antiserum Group B
Antiserum Group C
Antiserum Group D
Antiserum Group W135
Antiserum Group X
Antiserum Group Y
Antiserum Group Z
Antiserum Group Z prime (Z' )
Antiserum Poly 2, Groups X, Y, Z
Antiserum Poly, Groups A, B, C, D
Unit Size:
1mL
5mL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Description unitSize
11758333 1 mL Antiserum Group D 1mL
11762734 1 mL Antiserum Group Z prime (Z' ) 1mL
11748333 1 mL Antiserum Group C 1mL
11738333 1 mL Antiserum Group B 1mL
11728333 1 mL Antiserum Group A 1mL
11768333 1 mL Antiserum Poly, Groups A, B, C, D 1mL
11782734 1 mL Antiserum Group W135 1mL
11702904 1 mL Antiserum Group X 5mL
11712904 1 mL Antiserum Group Y 1mL
11772904 3 mL Antiserum Poly 2, Groups X, Y, Z 5mL
11742904 3 mL Antiserum Group Z 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code. 11758333 Supplier BD Supplier No. 222311
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
For serotyping Neisseria meningitidis in slide agglutination tests
  • Stable, lyophilized, polyclonal rabbit antisera
  • Contains 0.1% sodium azide as preservative
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance For in vitro diagnostic use
Description Antiserum Group D
Product Type Neisseria Meningitidis Antisera
Quantity 1 mL
For in vitro diagnostic use.