Learn More
BD Difco™ Legionella Agar Enrichment
Description
Applications:
Used with Legionella Agar base for isolation of Legionella
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | 2°C to 8°C |
| Additive | Legionella Agar Enrichment |
| Form | Liquid |
| For Use With (Application) | Legionella Agar base for the isolation of Legionella |
| Packaging Type | Bottle |
| Quantity | 5 mL |