Learn More
BD Difco™ Microbial Content Test Agar
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
Description
Certifications
CCAM
Specifications
Specifications
| Certifications/Compliance | CCAM |
| Form | Dehydrated |
| Media Type | Microbial Content Test Agar |
| For Use With (Application) | Detection and enumeration of microorganisms |
| Packaging Type | Pail |
| Quantity | 2 kg |