missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Terrific Broth
Click to view available options
:
Terrific Broth; 500g
Description
Terrific Broth is a highly enriched medium developed by Tartoff and Hobbs to improve yield in plasmid-bearing E. coli.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Terrific Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 12g/L, Yeast extract 24g/L, Dipotassium Phosphate 9.4g/L, Monopotassium Phosphate 2.2g/L |
| For Use With (Application) | Isolation of Escherichia coli |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction