missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Select APS™ Super Broth

Product Code. 17770467
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 17770467

Brand: BD 212485

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

To grow Escherichia coli to a high cell density

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage Refrigerate
Form Dehydrated
Media Type Super Broth Base, Select APSI (Alternative Protein Source)
Product Type Microbiology Media
Formulation Soy Hydrolysate 12g/L, Yeast extract 24g/L, Dipotassium Phosphate 11.4g/L, Monopotassium Phosphate 1.7g/L
For Use With (Application) Molecular Genetics medium used to grow Escherichia coli to a high cell density
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.