missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Sabouraud Brain Heart Infusion Agar Base
Use with chloromycetin and blood (optional) in cultivation and isolation of pathogenic fungi
Brand: BD 279720
Additional Details : Weight : 0.80000kg
Specifications
<30°C | |
SABHI Agar Base | |
Brain heart digest 9.25g/L, Proteose Peptone 5g/L, Enzymatic digest of Casein 5g/L, Dextrose 21g/L, Sodium Chloride 2.5g/L, Disodium Phosphate 1.25g/L, Agar 15g/L | |
Bottle |
Dehydrated | |
Agar | |
Cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and nonclinical specimens | |
500 g |