Learn More
BD Difco™ Dehydrated Culture Media: Rogosa Selective Lactobacillus (SL) Agar
Product Code. 11738942
Click to view available options
:
Rogosa Selective Lactobacillus (SL) Agar; 500g
Product Code. 11738942
Brand: BD 248020
For cultivating oral, vaginal and fecal lactobacilli
  • Tryptone 10g/L, Yeast extract 5g/L, Dextrose 10g/L, Arabinose 5g/L, Saccharose 5g/L, Sodium Acetate 15g/L, Ammonium Citrate 2g/L, Monopotassium Phosphate 6g/L, Magnesium Sulfate 0.57g Manganese Sulfate 0.12g Ferrous Sulfate 0.03g/L, Polysorbate 80 1g/L
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
COMPF
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance COMPF
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Rogosa Selective Lactobacillus Agar
Product Type Agar
Formulation Tryptone 10g/L, Yeast extract 5g/L, Dextrose 10g/L, Arabinose 5g/L, Saccharose 5g/L, Sodium Acetate 15g/L, Ammonium Citrate 2g/L, Monopotassium Phosphate 6g/L, Magnesium Sulfate 0.57g Manganese Sulfate 0.12g Ferrous Sulfate 0.03g/L, Polysorbate 80 1g/L
For Use With (Application) Selective cultivation of oral, vaginal and fecal lactobacilli
Packaging Type Bottle
Quantity 500 g