Learn More
BD Difco™ Dehydrated Culture Media: Modified Letheen Broth (AOAC)
Description
Applications:
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
Specifications
| Certifications/Compliance | BAM |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Letheen Broth |
| Product Type | Microbiology Media |
| Formulation | Letheen broth 25.7g/L, Tryptone 5g/L, Proteose Peptone No. 3 10g/L, Yeast extract 2g/L, Sodium Bisulfite 0.1g/L |
| For Use With (Application) | Microbiological evaluation of cosmetics - inactivates preservative agents |
| Packaging Type | Bottle |
| Quantity | 500 g |