Learn More
BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani)
Product Code. 11778902
Change view
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type Unit Size
11778902 500 g Bottle 500g
11768902 2000 g Pail 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11778902 Supplier BD Supplier No. 244620
  • ONLINE EXCLUSIVE PRICE
    955.00 NOK
    754.30 NOK / 500g
    Save 200.70 NOK
    21% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Propagate and maintain E. coli for molecular biology. BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
  • Contains 10g/L NaCl
  • For propagation of E. coli
  • Wide range of molecular biology applications
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CCAM
Content And Storage Refrigerate
Form Dehydrated
Media Type LB Broth, Miller
Product Type Microbiology Media
Formulation Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application) Propagation and maintenance of Escherichia coli for molecular biology
Packaging Type Bottle
Quantity 500 g