Learn More
BD Difco™ Dehydrated Culture Media: D-Mannitol
Click to view available options
:
D-Mannitol; BD Difco; 500g
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | <30°C |
| Additive | D-Mannitol |
| Form | Powder |
| For Use With (Application) | Dehydrated culture media |
| Packaging Type | Bottle |
| Quantity | 500 g |