Learn More
BD Difco™ Dehydrated Culture Media: Antibiotic Medium 9
Click to view available options
:
Antibiotic Medium 9 (Polymyxin Base Agar); 500g
Description
- Penassay Seed Agar Microbiological assay of antibiotics
Specifications
Specifications
| Certifications/Compliance | EP, USP |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Antibiotic Medium 9 |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 17g/L, Soy Peptone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Agar 20g/L |
| For Use With (Application) | Determination of antibiotic potency by the microbiological assay technique |
| Packaging Type | Bottle |
| Quantity | 500 g |