Learn More
BD Difco™ Dehydrated Culture Media: Anaerobic Agar
Click to view available options
:
Anaerobic Agar; 500g
Specifications
Specifications
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Anaerobic Agar |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 20g/L, Sodium Chloride 5g/L, Dextrose 10g/L, Agar 20g/L, Sodium Thioglycollate 2g/L, Sodium Formaldehyde Sulfoxylate 1g/L, Methylene blue 2mg/L |
| For Use With (Application) | For anaerobic bacteria |
| Packaging Type | Bottle |
| Quantity | 500 g |