missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Dehydrated Media Endo Agar
Description
- Danger of inhalation reduced with dehydration
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Product Type | Agar |
| Formulation | Dipotassium Phosphate 3.5g/L, Peptic digest of Animal Tissue 10g/L, Agar 15g/L, Lactose 10g/L, Sodium Sulfite 2.5g/L, Basic Fuchsin 0.5g/L |
| For Use With (Application) | Detection of Coliform other enteric microorganisms |
| Packaging Type | Bottle |
| Quantity | 500 g |
Safety and Handling
Signal Word
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P201-Obtain special instructions before use.,Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.,Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.,IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.,Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.,Dispose of contents/container to in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: sodium sulphite, basic fuchsin
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?