Learn More
Cytiva BBL™ Taxo™ Carbohydrate Discs: Trehalose
Description
- 0.5 in. discs impregnated with approximately 20mg of carbohydrate per discs
- Alternative to preparation and storage of carbohydrate solutions or media
Specifications
Specifications
| Antimicrobial Agents | Trehalose (TR) |
| Concentration | 20 mg |
| Packaging Type | Disc |
| Quantity | Each |