Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Vancomycin (30mcg)
Product Code. p-4898041
Click to view available options
Quantity:
10 Cartridges
Unit Size:
Pack of 10
Product Code. 17332381
Brand: BD 231353
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Vancomycin
Concentration 30 μg
Packaging Type Disc
Quantity 10 Cartridges