Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Bacitracin, B-2
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
- Employs Bauer-Kirby disc diffusion testing method
- Use in self-tamping dispensers
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
- Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
Specifications
| Antimicrobial Agents | Bacitracin |
| Concentration | 2U |
| Packaging Type | Disc |
| Quantity | 10 Cartridges |