Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Meropenem45
Product Code. p-4639646
Change view
Click to view available options
Quantity:
10 Pack
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17382401 10 Pack Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17382401 Supplier BD Supplier No. 231704
Add to basket
Add to basket
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Meropenem 45
Concentration 10 μg
No. per Pack 50
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use