Learn More
BD BBL™ Prepared RODAC™ and Contact Plate for Critical Environments
Product Code. 10792414
Change view
Click to view available options
:
Trypticase Soy Agar
Product Code.
10792414 Trypticase Soy Agar
1 options
Product Code. 10792414
Brand: BD 221288
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Recommended for Aerobic Plate Count (Microbial Test Limit) for water-miscible cosmetics containing preservatives
  • Trypticase™ Soy Agar with Lecithin and Polysorbate 80 (RODAC™).
      Designed for surface sampling for microbial contamination
    • Contains Trypticase™ Soy Agar with Lecithin and Polysorbate 80
    • Marked grids facilitate counting
    • Inward sloping walls and convex bottom keep agar bed in place during transit and use
Order Info
20 per sleeve
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Color Beige
Form Solid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar with Lecithin and Polysorbat
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
For Use With (Application) Detection and enumeration of microorganisms present on surfaces of sanitary importance
Packaging Type Plate
Quantity 20/Pk.