Learn More
BD BBL™ Prepared Plated Media: Clostridium difficile Selective Agar
Product Code. 17392351
Change view
Click to view available options
Clostridium difficile selective agar; 10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17392351 Clostridium difficile selective agar; 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17392351 Supplier BD Supplier No. 222228
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Selective and differential media for the primary isolation of Clostridium difficile from fecal specimens
  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce effects of drying during prolonged incubation
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Color Beige
Form Solid
Manufacturing Origin United States
Media Type Clostridium difficile selective Agar
Formulation Peptic digest of Animal Tissue 32g/L, Mannitol 6g/L, Monopotassium Phosphate 1g/L, Disodium Phosphate 5g/L, Sodium Chloride 2g/L, Growth Factors 3.3g/L, Magnesium Sulfate 0.1g/L, Agar 20g/L, Neutral red 0.03g/L, Cycloserine 0.25g/L, Cefoxitin 0.016g/L
For Use With (Application) Clostridium difficile from fecal specimens
Packaging Type Plate
Quantity 10/Pk.