Learn More
BD BBL™ Prepared Plated Media: CDC Anaerobe Laked Sheep Blood Agar w/Kanamycin and Vancomycin (KV)
Click to view available options
:
CDC Anaerobe Laked Sheep Blood Agar wiith Kanamycin and Vancomycin (KV); 20/Pk.
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Dark Red |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | CDC anaerobe Laked Sheep Blood Agar with Kanamycin and Vancomycin |
| Formulation | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 20g/L, Yeast extract 5g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, L-Cystine 0.4g/L, Kanamycin 0.1g/L, Vancomycin 0.0075g/L, Sheep Blood, defibrinated, laked 5% |
| For Use With (Application) | Fastidious and slow growing obligately anaerobic gram negative bacteria |
| Packaging Type | Plate |
| Quantity | 20/Pk. |