missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Urea Agar Slants, Complete (Christensen) Prepared Media

Product Code. 17342321
Change view
Click to view available options
Quantity:
10/Pk.
100 Pack
Unit Size:
Pack of 10
Pack of 100
Product Code. Quantity unitSize
17342321 10/Pk. Pack of 10
16411531 100 Pack Pack of 100
2 options
This item is not returnable. View return policy

Product Code. 17342321

Brand: BD 221096

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for the differentiation of organisms, especially the Enterobacteriaceae, on the basis of urease production

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM, ISO, USDA
Color Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Urea Agar
Formulation Pancreatic digest of Gelatin 1g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Potassium Phosphate 2g/L, Urea 20g/L, Phenol red 0.012g/L, Agar 15g/L
For Use With (Application) For differentiation of organisms, especially Enterobacteriaceae, on basis of Urease production
Packaging Type Glass Tube with Cap
Quantity 10/Pk.

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.