Learn More
BD BBL™ MGIT™ Mycobacteria Growth Indicator Tube: Reagents
Product Code. 17281352
Change view
Click to view available options
Unit Size:
1 set
Pack of 6
Product Code. unitSize
17281352 1 set
11766924 Pack of 6
2 options
Product Code. 17281352
Brand: BD 245116
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offers superior detection time in a user-friendly, fluorescent test format
  • Creates an environment suitable for rapid mycobacterial growth
  • Recommended for testing sputum, stool specimens, body fluids and tissue
  • The tube uses fluorometric technology to aid in the reading of results
  • Under UV light, a positive test emits a vivid orange fluorescent glow at the tube base and at the meniscus; a negative test shows little or no glow
  • The Mycobacteria can be removed from the tube for further testing
  • To use, supplement tube with MGIT OADC Enrichment and MGIT PANTA™ Antibiotic Mixture, inoculate with the prepared specimen and incubate
  • Tubes fit into standard procedures for decontamination and waste disposal
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE
Content And Storage 2 to 25°C
For Use With (Application) For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue
Quantity 6 Vials