Learn More
BD BBL™/Difco™ TB Brilliant Green K
Click to view available options
Quantity:
4 x 250 mL
Unit Size:
1 L
Description
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
Specifications
Specifications
| Type | TB Brilliant Green K |
| Quantity | 4 x 250 mL |
| For Use With (Application) | For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures |
| Storage Requirements | 15° to 30°C |
| Packaging Type | Bottle |
| Stain Type | Brilliant Green K |
For in vitro Diagnostic Use.