Learn More
BD BBL™ Dehydrated Culture Media: Trypticase Soy Agar (TSA II)
Product Code. 13277489
Change view
Click to view available options
:
Trypticase Soy Agar (TSA II); 500g
Product Code.
13277489 Trypticase Soy Agar (TSA II); 500g
1 options
Product Code. 13277489
Brand: BD 212305
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Supplementad with blood is used for cultivating fastidious microorganisms and for hemolytic reactions produced by many bacterial species
Certifications
BAM, COMPF
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance BAM, COMPF
Content And Storage 2 to 25°C
Form Dehydrated
Media Type Trypticase Soy Agar (TSA II)
Product Type Agar
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L
For Use With (Application) Cultivation of fastidious microorganisms and For the visualization of hemolytic reactions
Packaging Type Tube
Quantity 500 g