missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Regan-Lowe Charcoal Agar Base

Product Code. 15391546
Click to view available options
:
Regan-Lowe Charcoal Agar Base; 500g
This item is not returnable. View return policy

Product Code. 15391546

Brand: BD 298123

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For isolation of Bordetella pertussis from clinical specimens

  • Formulation: Beef extract 10g/L, Pancreatic digest of Casein 10g/L, Soluble Starch 10g/L, Sodium Chloride 5g/L, Charcoal 4g/L, Niacin 0.01g/L, Agar 12g/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

ISO/DIS, 11014

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 25°C
Form Dehydrated
Media Type Regan-Lowe Charcoal Agar Base
Product Type Agar
Formulation Beef extract 10g/L, Pancreatic digest of Casein 10g/L, Soluble Starch 10g/L, Sodium Chloride 5g/L, Charcoal 4g/L, Niacin 0.01g/L, Agar 12g/L
For Use With (Application) Isolation of Bordetella pertussis
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.