Learn More
BD BBL™ Dehydrated Culture Media: Mannitol Salt Agar
Description
Certifications
BAM, BS12, CMPH2, EP, JP, MCM9, USP
Specifications
Specifications
| Certifications/Compliance | BAM, BS12, CMPH2, EP, JP, MCM9, USP |
| Form | Dehydrated |
| Media Type | Mannitol Salt Agar |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Beef extract 1g/L, Sodium Chloride 75g/L, D-Mannitol 10g/L, Phenol red 25mg/L, Agar 15g/L |
| For Use With (Application) | For selective isolation and enumeration of staphylococci from clinical and nonclinical materials |
| Packaging Type | Bottle |
| Quantity | 500 g |