missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: MacConkey Agar without Crystal Violet

Product Code. 17302301
Change view
Click to view available options
Quantity:
500 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17302301 500 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17302301 Supplier BD Supplier No. 211393

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Less selective than MacConkey Agar, to permit growth of staphylococci and enterococci

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type MacConkey Agar
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Lactose 10g/L, Bile Salts 5g/L, Sodium Chloride 5g/L, Agar 12g/L, Neutral red 0.05g/L
For Use With (Application) Detection and isolation of gram negative organisms from clinical,1-3 dairy, 4 food, 5-7 Water, 8 pharmaceutical, 9-11 cosmetic, 6, 7 and other industrial source
Packaging Type Bottle
Quantity 500 g
compliance-icons
Product Identifier
  • MacConkey Agar w/o Crystal Violet
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: bile extract, 3-Trimethyl-2,8-phenazinediamine monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.