Description
- Cleanroom glove liners enhance operator comfort.
- Greatly reduce perspiration and contact dermatitis associated with wearing latex, urethane, PVC, nitrile and other chemical handling gloves.
- Liners can be repeatedly laundered and reused.
- Choose from a wide range of sizes that are color-coded for easy stock control.
- Key Atrributes
- 100% continuous filament knitted polyester
- Polyester offers a thinner yet durable barrier for the wearer.
- Full-finger design allows total protection for the most sensitive glove users
- Reusable, ambidextrous, seamless construction
- Applications
- For use in laboratories and manufacturing environments
- Can be used alone as inspection gloves to prevent fingerprints on circuit boards, glassware, lenses, optics and other critical surface
- Nylon glove liners provide a thin barrier for wet and temperature related processing
- The glove liner is suitable in any ISO class cleanroom when worn under a nitrile, latex, vinyl, neoprene or other type glove
Specifications
Specifications
| Construction | Seamless Knit |
| Coating Material | Uncoated |
| Color | White |
| Disposable | Reusable |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Polyester |
| Product Type | Glove Liners |
| Size | Large |
| Sterility | Non-sterile |
| Show More |
Product Title
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?