Learn More
Abnova™ BCL6 Split FISH Probe
Product Code. 16033525
Change view
Click to view available options
Quantity:
100 uL
200 uL
Unit Size:
100µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16033525 200 uL 200µL
16023525 100 uL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16033525 Supplier Abnova™ Supplier No. FS0068.200uL
  • ONLINE EXCLUSIVE PRICE
    33840.00 NOK
    32466.87 NOK / 200µL
    Save 1373.13 NOK
    4% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Includes
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Specifications
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application) FISH-Ce
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Quantity 200 uL