missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-3 Monoclonal Antibody (Ham 150-3.5), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Thermo Scientific MA517668
Additional Details : Weight : 0.01000kg
Description
B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.Specifications
Bcl-3 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Q9Z2F6 | |
Bcl3 | |
recombinant Bcl-3-6-histidine tag | |
300 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 λ |
Flow Cytometry | |
Ham 150-3.5 | |
FITC | |
Bcl3 | |
AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3 | |
Armenian Hamster | |
Protein G | |
RUO | |
12051 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |