missing translation for 'onlineSavingsMsg'
Learn More

BCAR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13628-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16891934

  • 3468.15 NOK / 150µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Breast tumors are initially dependent on estrogens for growth and progression and can be inhibited by anti-estrogens such as tamoxifen. However, breast cancers progress to become anti-estrogen resistant. Breast cancer anti-estrogen resistance gene 3 was identified in the search for genes involved in the development of estrogen resistance. The gene encodes a component of intracellular signal transduction that causes estrogen-independent proliferation in human breast cancer cells. The protein contains a putative src homology 2 (SH2) domain, a hall mark of cellular tyrosine kinase signaling molecules, and is partly homologous to the cell division cycle protein CDC48.
Specifications

Specifications

BCAR3
Polyclonal
Unconjugated
BCAR3
BCAR3, KIAA0554, Novel SH2 containing protein 2, NSP2, SH2D3B
Rabbit
Antigen Affinity Chromatography
RUO
29815, 310838, 8412
-20°C
Liquid
Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75815, Q9QZK2
BCAR3
BCAR3 Fusion Protein Ag4546
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers