Learn More
BBT594, MedChemExpress
Product Code. 16483647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483647 50 mg 50mg
16467638 10 mg 10mg
16447546 100 mg 100mg
16440108 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16483647 Supplier Medchem Express Supplier No. HY18840
MedChemExpress BBT594 is a potent receptor tyrosine kinase RET inhibitor, used for cancer treatment.
BBT594 is a potent receptor tyrosine kinase RET inhibitor, used for cancer treatment.
Specifications
Chemical Name or Material BBT594
CAS 882405-89-2
Color White
Purity Grade Notes Research
Molecular Formula C28H30F3N7O3
Quantity 50 mg
Synonym NVP-BBT594
Solubility Information DMSO : ≥ 33 mg/mL (57.94 mM)
SMILES O=C(N1CCC2=C1C=CC(OC3=NC=NC(NC(C)=O)=C3)=C2)NC4=CC=C(CN5CCN(C)CC5)C(C(F)(F)F)=C4
Molecular Weight (g/mol) 569.58
Formula Weight 569.58
Percent Purity 96.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease