Learn More
BBS8 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12505-1-AP-20UL
 View more versions of this product
Product Code. 16850254
  • 1710.33 NOK / 20µL
Description
The BBS8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BBS8. This antibody recognizes human, mouse, rat antigen. The BBS8 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
BBS8
Polyclonal
Unconjugated
20 μL
123016
IgG
Western Blot, Immunoprecipitation, ELISA
353 μg/mL
Rabbit
Primary
Human, Rat, Mouse