Learn More
BBS8 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12505-1-AP-20UL
Description
The BBS8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BBS8. This antibody recognizes human, mouse, rat antigen. The BBS8 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| BBS8 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 123016 | |
| IgG |
| Western Blot, Immunoprecipitation, ELISA | |
| 353 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |