missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18961-1-AP-20UL
This item is not returnable.
View return policy
Description
BBS7, also named as BBS2L1, is a component of the BBSome complex which is required for ciliogenesis but is dispensable for centriolar satellite function. BBS7 is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome.Specifications
| BBS7 | |
| Polyclonal | |
| Unconjugated | |
| BBS7 | |
| Bardet Biedl syndrome 7, BBS2 like protein 1, BBS2L1, BBS7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55212, 71492 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IWZ6, Q8K2G4 | |
| BBS7 | |
| BBS7 Fusion Protein Ag4601 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto