Learn More
BBS5 Mouse anti-Human, Mouse, Clone: 4G8E12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66136-1-IG-150UL
This item is not returnable.
View return policy
Description
The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. The BBSome complex, together with the LTZL1, controls SMO ciliary trafficking and contributes to the sonic hedgehog (SHH) pathway regulation. Required for BBSome complex ciliary localization but not for the proper complex assembly.Specifications
| BBS5 | |
| Monoclonal | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N3I7, Q9CZQ9 | |
| Bbs5 | |
| BBS5 Fusion Protein Ag6844 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4G8E12 | |
| Unconjugated | |
| BBS5 | |
| Bardet Biedl syndrome 5, BBS5 | |
| Mouse | |
| Protein G | |
| RUO | |
| 129880, 72569 | |
| -20°C | |
| Liquid |