missing translation for 'onlineSavingsMsg'
Learn More

BBS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16850664 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16850664 20 μL 20µL
16840664 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16850664

Brand: Proteintech 127661AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BBS4 contains tetratricopeptide repeats (TPR), similar to O-linked N-acetyglucosamine transferase. Mutations in the gene encoding this protein have been observed in patients with Bardet-Biedl syndrome type 4. BBS4 may play a role in pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation.

Specifications

Antigen BBS4
Applications Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BBS4
Gene Accession No. Q8C1Z7, Q96RK4
Gene Alias Bardet Biedl syndrome 4, BBS4
Gene Symbols BBS4
Host Species Rabbit
Immunogen BBS4 Fusion Protein Ag3430
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102774, 300754, 585
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.