missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS10 Mouse anti-Human, Clone: 4C6A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66679-1-IG-20UL
This item is not returnable.
View return policy
Description
BBS10 is a member of the Bardet-Biedl syndrome (BBS) gene family. Bardet-Biedl syndrome is an autosomal recessive disorder characterized by progressive retinal degeneration, obesity, polydactyly, renal malformation and mental retardation. The proteins encoded by BBS gene family members are structurally diverse and the similar phenotypes exhibited by mutations in BBS gene family members is likely due to their shared roles in cilia formation and function. Many BBS proteins localize to the basal bodies, ciliary axonemes, and pericentriolar regions of cells. BBS proteins may also be involved in intracellular trafficking via microtubule-related transport. The protein encoded by this gene is likely not a ciliary protein but rather has distant sequence homology to type II chaperonins. As a molecular chaperone, this protein may affect the folding or stability of other ciliary or basal body proteins. Inhibition of this protein′s expression impairs ciliogenesis in preadipocytes.Specifications
| BBS10 | |
| Monoclonal | |
| 0.95 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8TAM1 | |
| BBS10 | |
| BBS10 Fusion Protein Ag17960 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4C6A10 | |
| Unconjugated | |
| BBS10 | |
| Bardet Biedl syndrome 10, BBS10, C12orf58, FLJ23560 | |
| Mouse | |
| Protein A | |
| RUO | |
| 79738 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction