Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Chloramphenicol29
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
| Antimicrobial Agents | Chloramphenicol29 |
| Concentration | 30 μg |
| Packaging Type | Disc |
| Quantity | 10 Pack |
For in vitro diagnostic use