Learn More
BD BBL™ Prepared Lowenstein-Jensen Media
Product Code. 11938536
Change view
Click to view available options
Quantity:
100/Pk.
Packaging Type:
Slants (A tubes)
Unit Size:
Pack of 100
Product Code. Quantity Packaging Type unitSize
11938536 100/Pk. Slants (A tubes) Pack of 100
1 options
Product Code. 11938536
Brand: BD 220909
Used for cultivation of Mycobacterium tuberculosis and other mycobacterial species
  • Inspissated, egg-based media
  • Available as Mycoflask™ bottles, prepared slants and bottles
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE, BS12, CMPH2, EP, MCM9, SMWW
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Lowenstein-Jensen Media
Formulation Monopotassium Phosphate 2.5g/L, Magnesium Sulfate 0.24g/L, Sodium Citrate 0.6g/L, L-Asparagine 3.6g/L, Potato Flour 30g/L, Malachitegreen 0.4g/L, Glycerol 12mL, Whole Egg 1000mL
For Use With (Application) Cultivation of Mycobacterium tuberculosis and other mycobacterial species
Packaging Type Slants (A tubes)
Quantity 100/Pk.
For in vitro diagnostic use.