Learn More
BAY-218, MedChemExpress
Product Code. 16490767
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16490767 50 mg 50mg
16487177 5 mg 5mg
16485336 100 mg 100mg
16444458 10 mg 10mg
16423096 25 mg 25mg
5 options
Product Code. 16490767
Brand: Medchem Express HY111449
MedChemExpress BAY-218 (AHR antagonist 1) is an aryl hydrocarbon receptor (AHR) antagonist extracted from patent WO2017202816A1, example 23, has an IC50 of 39.9 nM in human cell line.
BAY-218 (AHR antagonist 1) is an aryl hydrocarbon receptor (AHR) antagonist extracted from patent WO2017202816A1, example 23, has an IC50 of 39.9 nM in human cell line.
Specifications
Chemical Name or Material BAY-218
CAS 2162982-11-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H17ClFN3O3
Quantity 50 mg
Synonym AHR antagonist 1
Solubility Information DMSO : ≥ 250 mg/mL (622.17 mM)
SMILES O=C(C1=CC(C2=CC=C(Cl)C=C2)=NN(C3=CC=CC(F)=C3)C1=O)N[C@@H](C)CO
Molecular Weight (g/mol) 401.82
Formula Weight 401.82
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death