missing translation for 'onlineSavingsMsg'
Learn More

BAY-218, MedChemExpress

Product Code. 16444458
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444458

Brand: Medchem Express HY111449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY-218 (AHR antagonist 1) is an aryl hydrocarbon receptor (AHR) antagonist extracted from patent WO2017202816A1, example 23, has an IC50 of 39.9 nM in human cell line.

BAY-218 (AHR antagonist 1) is an aryl hydrocarbon receptor (AHR) antagonist extracted from patent WO2017202816A1, example 23, has an IC50 of 39.9 nM in human cell line.

Specifications

Chemical Name or Material BAY-218
CAS 2162982-11-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H17ClFN3O3
Quantity 10 mg
Synonym AHR antagonist 1
Solubility Information DMSO : ≥ 250 mg/mL (622.17 mM)
SMILES O=C(C1=CC(C2=CC=C(Cl)C=C2)=NN(C3=CC=CC(F)=C3)C1=O)N[C@@H](C)CO
Molecular Weight (g/mol) 401.82
Formula Weight 401.82
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.