missing translation for 'onlineSavingsMsg'
Learn More

BAY-218, MedChemExpress

Product Code. 16423096
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423096 25 mg 25mg
16490767 50 mg 50mg
16487177 5 mg 5mg
16485336 100 mg 100mg
16444458 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16423096 Supplier Medchem Express Supplier No. HY111449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY-218 (AHR antagonist 1) is an aryl hydrocarbon receptor (AHR) antagonist extracted from patent WO2017202816A1, example 23, has an IC50 of 39.9 nM in human cell line.

BAY-218 (AHR antagonist 1) is an aryl hydrocarbon receptor (AHR) antagonist extracted from patent WO2017202816A1, example 23, has an IC50 of 39.9 nM in human cell line.

Specifications

Chemical Name or Material BAY-218
CAS 2162982-11-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H17ClFN3O3
Quantity 25 mg
Synonym AHR antagonist 1
Solubility Information DMSO : ≥ 250 mg/mL (622.17 mM)
SMILES O=C(C1=CC(C2=CC=C(Cl)C=C2)=NN(C3=CC=CC(F)=C3)C1=O)N[C@@H](C)CO
Molecular Weight (g/mol) 401.82
Formula Weight 401.82
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.