Learn More
Thermo Scientific™ Oxoid™ Bax™ E.Coli 0157:H7 MP Medium
Click to view available options
Quantity:
2.5 kg
Unit Size:
3kg
Description
The Bax™ E.Coli 0157:H7 MP medium from Oxoid is a selective agar for the isolation and differentiation of enterohemorrhagic (EHEC) E. coli O157:H7 strains from food and clinical material.
Specifications
Specifications
| Product Type | Microbiology Media |
| For Use With (Application) | For the isolation and differentiation of enterohemorrhagic E. coli O157:H7 strains from food and clinical material |
| Quantity | 2.5 kg |