Learn More
Asecos™ BATTERY STORE Storage Cabinet ION-CLASSIC-90
Product Code. 16389406 Shop All asecos Products
Click to view available options
No. of Doors:
1
2
No. of Shelves:
3
4
5
6
Unit Size:
Each
For indoor use only
Product Code. 16389406
Brand: Asecos™ 3725404737264
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For indoor use only
Safe passive storage of lithium-ion batteries
  • Type 90 Safety storage cabinet
  • Tall cabinet with self-closing wing doors
  • With cylinder locking and locking state indicator
  • Integrated adjusting aids to compensate uneven floor
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Integrated transport base with removable, optional base cover
  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • CE compliant
Specifications
Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, CE compliant
Dimensions (D x W x H) Interior 522 x 1050 x 1647 mm
Dimensions (D x W x H) 615 x 1193 x 2224 mm
Door Style Wing
For Use With (Application) Safe passive storage of lithium-ion batteries
Includes Bottom collecting sump (sheet steel powder-coated)
Model IO90.195.120.K1.WDC
No. of Doors 2
No. of Shelves 4
Shelf Type Grid
Surface Coating Powder-coated textured
Color Anthracite Gray, Gentian Blue
Material Sheet Steel
Product Type Storage cabinet