Learn More
Testo™ Batteries
Product Code. 10002772
Change view
Click to view available options
For Use With (Equipment):
Testo 175-T1, 175-T2, 177
Testo 175-T3, 175-H1, 175-H2, 175-S1, 175-S2
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10002772 Testo 175-T1, 175-T2, 177 Each
10192492 Testo 175-T3, 175-H1, 175-H2, 175-S1, 175-S2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10002772 Supplier Testo™ Supplier No. 0515 0177
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use this battery with a variety of products. Testo™ Batteries are compatible with Testo data loggers.
  • 3.6V
  • 0.8Ah
  • ½AA
Specifications
Product Type Battery
For Use With (Equipment) Testo 175-T1, 175-T2, 177
Voltage 3.6 V
Amperage 1.9 Ah