Learn More
Julabo™ Bath Adapters
Product Code. 15254263
Change view
Click to view available options
Size:
G1 x 1/4 in. F to BF 1/2 in. ID
M10 x 1 to M16 x 1
M16 x 1 F to NPT 1/2 in. F
M16 x 1 female to 2 x M16 x 1 male
M16 x 1 female to M16 x 1 female
M16 x 1 female to NPT 1/2 in. male
M16 x 1 female to NPT 1/4 in. male
M16 x 1 female to NPT 3/8 in. male
M16 x 1 female to tube 1/4 in. male
M16 x 1 female to tube 3/8 in. male
M16 x 1 male to NPT 1/4 in. female
M16 x 1 male/male
M24 x 1/2 F to NPT 1/2 in. F
M24 x 1/2 F to NPT 3/8 in. M
M24 x 1/2 in. F to M16 x 1 M
M24 x 1/2 male to M24 x 1/2 male
Unit Size:
Each
Pack of 2
Product Code. Size unitSize
15254263 M24 x 1/2 F to NPT 3/8 in. M Pack of 2
11974708 G1 x 1/4 in. F to BF 1/2 in. ID Pack of 2
10318503 M10 x 1 to M16 x 1 Each
11964718 M16 x 1 female to 2 x M16 x 1 male Each
11984688 M16 x 1 female to M16 x 1 female Pack of 2
11954708 M16 x 1 F to NPT 1/2 in. F Pack of 2
11904708 M16 x 1 female to NPT 1/2 in. male Pack of 2
11917458 M16 x 1 female to NPT 3/8 in. male Pack of 2
11954698 M16 x 1 female to tube 1/4 in. male Pack of 2
10052822 M16 x 1 male/male Each
13460269 M24 x 1/2 in. F to M16 x 1 M Pack of 2
16255472 M24 x 1/2 male to M24 x 1/2 male Each
11907458 M16 x 1 female to NPT 1/4 in. male Pack of 2
11944698 M16 x 1 male to NPT 1/4 in. female Pack of 2
11964698 M16 x 1 female to tube 3/8 in. male Pack of 2
15224263 M24 x 1/2 F to NPT 1/2 in. F Pack of 2
16 options
Product Code. 15254263
Brand: Julabo™ 8890059
Only null left
Specifications
For Use With (Equipment) Magnum 91, PRESTO™ A30, PRESTO™ A40, PRESTO™ A80, PRESTO™ W40 temperature control/process system
Type Adapter
Size M24 x 1/2 F to NPT 3/8 in. M